Learn More
Medchemexpress LLC 2-[4-({[2-(benzodioxol-5-yloxy)pyridin-3-yl]carbonyl}amino)methyl]-3-fluorophenoxypropionic acid | 445295-04-5 | MFCD31382154 | 100.0% | 454.40 g·mol⁻1 | C23H19FN2O7 | 1 ML

Supplier: Medchemexpress LLC HY10180310MM 1ML
CP671305 is a potent, orally active, selective inhibitor of phosphodiesterase-4-D provided for research use. It is offered as a solid and as a ready-to-use 10 mM solution in DMSO (1 mL), and is characterized by high reported purity and documented physicochemical properties suitable for in vitro and in vivo studies.
- Potent, orally active selective phosphodiesterase-4-D inhibitor.
- Available as solid and as 10 mM solution in DMSO (1 mL).
- High reported purity (99.95%).
- Molecular weight 454.40 g·mol⁻1; formula C23H19FN2O7.
- Soluble in DMSO (~100 mg/mL) with ultrasonic assistance.
- Recommended storage: powder and solution conditions as specified.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.