Learn More
Selleck Chemical LLC Apabetalone (RVX-208) 5mg 1044870-39-4 RVX-000222
SDP
Supplier:  Selleck Chemical LLC S72955MG
Encompass_Preferred
Apabetalone (RVX-208, RVX-000222) is a potent BET bromodomain inhibitor with IC50 of 0.510 ?M for BD2 in a cell-free assay, about 170-fold selectivity over BD1. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0668254