Learn More
Medchemexpress LLC 2-[4-[(2S)-4-[(6-amino-3-pyridinyl)sulfonyl]-2-prop-1-ynylpiperazin-1-yl]phenyl]-1,1,1,3,3,3-hexafluoropropan-2-ol | 1361224-53-4 | MFCD28386194 | 99.5% | 522.46 g/mol | C21H20F6N4O3S | 5MG

Supplier: Medchemexpress LLC HY124115MG
AMG-3969 is a small-molecule research compound that disrupts the interaction between glucokinase (GK) and glucokinase regulatory protein (GKRP). It is supplied for research use in in vitro and in vivo studies and is reported to have potent activity against the GK-GKRP interaction.
- Disrupts GK-GKRP interaction; reported IC50 = 4 nM.
- High purity: 99.45% (supplier-reported).
- Molecular formula: C21H20F6N4O3S; molecular weight: 522.46 g/mol.
- Soluble in DMSO.
- Suitable for in vitro and in vivo research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.