Learn More
Medchemexpress LLC 2-(4,5-dihydro-1,3-thiazol-2-ylsulfanyl)-1-(3,4-dihydroxyphenyl)ethanone hydrochloride | 1036069-26-7 | MFCD11114394 | 99.4% | 269.34 g/mol | C11H11NO3S2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1086365MG
Encompass_Preferred
RETRA is a small-molecule research compound that acts as a mutant p53-dependent activator of p73, increasing p73 expression and releasing p73 from inhibitory complexes to produce tumor-suppressor effects. It is supplied as a solid for laboratory use (CAS 1036069-26-7; molecular weight 269.34 g/mol).
  • Mutant p53-dependent activator of p73.
  • Increases p73 expression and releases it from inhibitory complexes.
  • High reported purity suitable for research (99.4%).
  • Supplied in small research pack sizes, including 5 mg.
  • Store at -20°C and protect from light; in solvent store at -80°C for long-term.
Catalog No. 50-002-63835