Learn More
Medchemexpress LLC 2,4,5-trimethoxybenzaldehyde | 4460-86-0 | MFCD00003312 | 99.8% | 196.20 g/mol | C10H12O4 | 25 G
SDP
Supplier:  Medchemexpress LLC HY10058025G
Encompass_Preferred
Asaraldehyde (2,4,5-trimethoxybenzaldehyde) is a small-molecule research reagent reported to inhibit cyclooxygenase-2 (COX-2). Supplied as an off-white to light brown solid with a high assay purity, it is intended for biochemical assays, analytical standards, and research applications where COX-2 modulation or reference material is required.
  • COX-2 inhibitory activity (IC50 100 μg/mL)
  • High purity suitable for research and analytical use
  • Solid form for straightforward storage and handling
  • Molecular formula C10H12O4; molecular weight 196.20 g/mol
  • Available in common laboratory pack sizes, including 25 G
Catalog No. 50-002-11878