Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,4,6-Tri-2-pyridinyl-1,3,5-triazine | 3682-35-7 | MFCD00006045 | 99.9% | 10 G
SDP

Supplier:  Medchemexpress LLC HYD022110G

Encompass_Preferred

2,4,6-Tri-2-pyridinyl-1,3,5-triazine (NSC 112125) is a colorimetric reagent used for detecting iron. It forms a complex with Fe (II) that can be quantified as a measure of iron concentration by colorimetric detection at 594 nm. This product is for research use only.

  • Purity: 99.87%
  • Molecular weight: 312.34
  • Formula: C18H12N6
  • CAS no.: 3682-35-7
  • Appearance: Solid
  • Color: White to yellow
  • Solubility: Methanol: 50 mg/mL, DMSO: 2 mg/mL

Catalog No. 50-004-10710


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.