Learn More
Medchemexpress LLC (2,4,6-Trimethyl-phenoxy)-acetic acid | 13333-81-8 | MFCD00224887 | 98.0% | 194.23 g/mol | C11H14O3 | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW264306250MG
Encompass_Preferred
2-(2,4,6-Trimethylphenoxy)acetic acid is an organic research chemical described as having herbicide activity. It is supplied in laboratory-scale quantities for research use. Key identifiers include CAS 13333-81-8, formula C11H14O3, molecular weight 194.23 g/mol, and typical purity 98%.
  • Organic compound reported to have herbicide activity.
  • Supplied in laboratory pack sizes for experimental use.
  • Typical high purity supports reproducible results.
  • Chemical formula C11H14O3 and molecular weight 194.23 g/mol for reference.
  • Product datasheet provides handling and safety information.
Catalog No. 50-002-81225