Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2,4,6-trimethyl-phenoxy)-acetic acid | 13333-81-8 | MFCD00224887 | 99.5% | 194.23 g/mol | C11H14O3 | 1 G
SDP

Supplier:  Medchemexpress LLC HYW2643061G

Encompass_Preferred

(2,4,6-Trimethyl-phenoxy)-acetic acid is a small-molecule herbicide research reagent provided as a powder for laboratory use. It is suitable for biochemical screening, synthesis, and herbicide development studies, and is supplied with analytical and safety documentation for quality control and safe handling.

  • High purity (99.5%).
  • Powder form, convenient for synthesis and assay preparation.
  • Available in multiple laboratory sizes, including 1 G.
  • Molecular formula C11H14O3; molecular weight 194.23 g/mol.
  • CAS 13333-81-8 for unambiguous identification.
  • Supplied with data sheet, COA, and SDS for quality and safety documentation.
  • Store powder at -20°C for long-term stability; follow SDS for solvent storage.

Catalog No. 50-002-81216


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.