Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2,4,6-trimethyl-phenoxy)-acetic acid | 13333-81-8 | MFCD00224887 | 99.5% | 194.23 g/mol | C11H14O3 | 5 G
SDP

Supplier:  Medchemexpress LLC HYW2643065G

Encompass_Preferred

2-(2,4,6-Trimethylphenoxy)acetic acid is a research-grade herbicidal small molecule supplied as a solid powder for laboratory use. The material is provided with reported high purity and characterized molecular data for analytical and preparative applications. Handle and store according to the supplier datasheet and safety guidance.

  • High purity (99.5%).
  • Solid powder suitable for bench-scale work.
  • Molecular weight 194.23 g/mol; chemical formula C11H14O3.
  • Available in small research sizes including 5 G.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years).
  • Intended for research use only; follow SDS handling instructions.

Catalog No. 50-002-81212


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.