Learn More
Medchemexpress LLC 2 4 6-Triphenylanili 250mg
SDP
Supplier:  Medchemexpress LLC 250MGHYW112651
Encompass_Preferred
2 4 6-Triphenylaniline has anti-diabetic activity and can be encapsulated in nano-emulsions (NE) to enhance stability and permeability The NE loaded with 2 4 6-Triphenylaniline inhibits -glucosidase and -amylase[1]
Catalog No. 50-002-04535