Learn More
Medchemexpress LLC [2-[4-amino-6-[(4-phenoxyphenyl)methylamino]-1,3,5-triazin-2-yl]-3-fluorophenyl]methanol | 2375070-79-2 | 99.9% | C23H20FN5O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13449410MG
Encompass_Preferred
MS48107 is a research-grade small molecule positive allosteric modulator of the proton-sensing G protein-coupled receptor GPR68. It is potent, selective, and reported to be brain-penetrant in mice, making it suitable as an in vitro and in vivo pharmacology tool when used with appropriate controls and experimental validation.
  • Potent and selective positive allosteric modulator of GPR68.
  • Demonstrated brain penetration in mouse models.
  • High reported purity (99.9%).
  • Available as solid and as ready-to-use DMSO solution.
  • Recommended cold storage for long-term stability.
Catalog No. 50-002-61530