Learn More
Medchemexpress LLC 2-(4-bromophenyl)-N-(4-fluorobenzyl)-3-propyl-3H-imidazo[4,5-b]pyridin-5-amine | 1397255-09-2 | MFCD32201091 | 99.6% | 439.32 | C22H20BrFN4 | 25MG
SDP
Supplier:  Medchemexpress LLC HY12622525MG
Encompass_Preferred
SJA710-6 is a small-molecule research reagent reported to selectively induce differentiation of mesenchymal stem cells (MSCs) toward hepatocyte-like cells. The compound has CAS 1397255-09-2, molecular formula C22H20BrFN4, molecular weight 439.32 g/mol, and is provided with high purity for in vitro research applications.
  • Selective inducer of MSC differentiation toward hepatocyte-like cells.
  • Molecular formula C22H20BrFN4 and molecular weight 439.32 g/mol.
  • High purity: 99.6%.
  • Available in multiple pack sizes suitable for small-scale research.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-34984