Learn More
Medchemexpress LLC 2-(4-chloro-2-methylphenoxy)propanoic acid | 93-65-2 | MFCD00002648 | 99.8% | 214.65 g/mol | C10H11ClO3 | 500 MG

Supplier: Medchemexpress LLC HY118126500MG
Mecoprop (2-(4-chloro-2-methylphenoxy)propanoic acid) is supplied as a high-purity analytical standard for research and analytical applications. The material is intended for use as a reference standard in method development, calibration, and analytical testing, and is provided in solid form with options for DMSO solutions.
- Purity 99.8%.
- CAS number 93-65-2.
- Molecular weight 214.65 g/mol.
- Chemical formula C10H11ClO3.
- Soluble in DMSO; ready-made DMSO solutions are available.
- Intended for research and analytical use; not for human or therapeutic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.