Learn More
Medchemexpress LLC 2-(4-chloro-2-methylphenoxy)propionic acid | 93-65-2 | MFCD00002648 | 99.8% | 214.65 | C10H11ClO3 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1181261G
Encompass_Preferred
Mecoprop is a chlorinated phenoxypropionic acid herbicide supplied as an analytical standard for research and analytical applications. It is available as a solid and as ready-to-use DMSO solutions for method development and quality-control assays.
  • High purity (99.8%), suitable for analytical use.
  • Chemical formula C10H11ClO3, molecular weight 214.65.
  • Supplied as a solid or as a 10 mM solution in DMSO for ready-to-use assays.
  • Intended for research, method development, and analytical applications.
  • Documentation available: data sheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-27768