Learn More
Medchemexpress LLC 2-(4-chlorophenoxy)ethanol | 1892-43-9 | MFCD00014036 | 97.3% | 172.61 g/mol | C8H9ClO2 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0168575G
Encompass_Preferred
2-(4-chlorophenoxy)ethanol is an aromatic ether used as a chemical intermediate in small-molecule synthesis, notably for herbicide development. It has the molecular formula C8H9ClO2, molecular weight 172.61 g/mol, and CAS 1892-43-9. The material is supplied for research use at high purity and is suitable for laboratory-scale synthetic work.
  • Used as an intermediate in herbicide and fine chemical synthesis.
  • High purity for reliable synthetic performance.
  • Soluble in DMSO, compatible with common organic reaction protocols.
  • Available in small packages suitable for lab-scale work.
  • Simple handling and storage under standard laboratory conditions.
Catalog No. 50-002-99986