Learn More
Medchemexpress LLC 2-((4-chlorophenyl)thio)-N-(4-(pyridin-2-yl)thiazol-2-yl)acetamide | 895470-67-4 | 99.7% | 361.87 | C16H12ClN3OS2 | 5MG
SDP
Supplier:  Medchemexpress LLC HY1248155MG
Encompass_Preferred
CP-312 is a small-molecule inducer of heme oxygenase-1 (HO-1) that activates the antioxidant response and protects human iPSC-derived cardiomyocytes from oxidative stress by inducing HMOX1 expression. Supplied for research use, it is provided as a solid with defined purity and storage recommendations.
  • Induces heme oxygenase-1 and activates antioxidant response.
  • Protects iPSC-derived cardiomyocytes from oxidative stress.
  • High purity (99.7%) suitable for research applications.
  • Solid, off-white to pink appearance for easy handling.
  • Stable under recommended storage conditions for prolonged use.
  • Available in multiple pack sizes for experimental flexibility.
Catalog No. 50-002-39303