Learn More
Medchemexpress LLC 2,4-d-13C6 (2,4-dichlorophenoxyacetic acid-13C6) | 150907-52-1 | 97.5% | 226.99 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY18572S1MG
Encompass_Preferred
2,4-D-13C6 is the 13C-labeled version of 2,4-Dichlorophenoxyacetic acid, a selective systemic herbicide. It controls broad-leaved weeds by functioning as a plant hormone, inducing uncontrolled growth in meristematic tissues, and inhibiting DNA and protein synthesis. Stable heavy isotopes, such as carbon-13, are incorporated into the molecule for use as tracers in quantitative analysis during drug development.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Selective systemic herbicide.
  • Acts as a plant hormone.
  • Inhibits DNA and protein synthesis.
Catalog No. 50-003-87785