Learn More
Medchemexpress LLC 2,4-D (2,4-dichlorophenoxyacetic acid) | 94-75-7 | 99.9% | 221.04 | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HY1857210MM/1ML
Encompass_Preferred
2,4-D (2,4-Dichlorophenoxyacetic acid) is a selective herbicide used for the control of broad-leaved weeds. It can induce apoptosis and inhibits DNA and protein synthesis, thereby preventing normal plant growth and development.
  • Functions as a selective herbicide.
  • Controls broad-leaved weeds.
  • Induces apoptosis.
  • Inhibits DNA and protein synthesis.
  • Prevents normal plant growth and development.
  • High purity for research applications.
Catalog No. 50-003-87819