Learn More
Medchemexpress LLC 2,4-D sodium | 2702-72-9 | ≥98.0% | 243.02 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY18572A500MG
Encompass_Preferred
2,4-D sodium (2,4-Dichlorophenoxyacetate) is a selective herbicide effective against broad-leaved weeds. It functions by inducing apoptosis and inhibiting DNA and protein synthesis, thereby disrupting normal plant growth and development. In laboratory studies, it inhibits the viability of A549 and WI38 cells with IC50 values of 126 μM and 115 μM, respectively, and can induce apoptosis by influencing the cell cycle. In vivo studies have also shown toxic effects on the reproductive system of male rats, including reduced organ weights and sperm viability.
  • Selective herbicide for broad-leaved weeds.
  • Induces apoptosis and inhibits DNA and protein synthesis.
  • Inhibits A549 and WI38 cell viability in vitro.
  • Affects cell cycle and induces apoptosis.
  • Causes reproductive toxicity in male rats.
  • Reduces organ weights and sperm viability in vivo.
  • Increases FSH and LH levels in vivo.
Catalog No. 50-003-88001