Learn More
Medchemexpress LLC 2,4-decadienal | 2363-88-4 | MFCD00007007 | 95.1% | 152.23 g/mol | C10H16O | 1mL
SDP
Supplier:  Medchemexpress LLC HYW76073310MM/1ML
Encompass_Preferred
2 4-Decadienal is a toxic aldehyde produced by the oxidation of linoleic acid-rich oils 2 4-decadienal can be used as an alternative oxidation indicator for linoleic acid-rich oils[1]
Catalog No. 50-002-69509