Learn More
Medchemexpress LLC 2,4-Dibromophenol-d3 | 1219805-97-6 | 99.5% | 254.92 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW009490S10MG
Encompass_Preferred
2,4-Dibromophenol-d3 is the deuterium labeled 2,4-Dibromophenol. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis
Catalog No. 50-004-53097