Learn More
Medchemexpress LLC 2,4-Dibromophenol-d3 | 1219805-97-6 | 99.5% | 254.92 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW009490S5MG
Encompass_Preferred
2,4-Dibromophenol-d3 is the deuterium-labeled form of 2,4-Dibromophenol. Stable heavy isotopes of elements such as hydrogen and carbon are incorporated into drug molecules primarily to serve as tracers for quantitation during the drug development process. Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-66593