Learn More
Medchemexpress LLC 2,4-dichloro-3,5-dimethylphenol | 133-53-9 | MFCD00019981 | 95.4% | 191.05 g/mol | C8H8Cl2O | 1 KG
SDP
Supplier:  Medchemexpress LLC HYW1407991KG
Encompass_Preferred
Dichloroxylenol is a chlorinated phenolic compound used as a bactericide, preservative, and deodorant for laboratory research. It is applied in microbiological and veterinary studies, including research into uterine infections in cattle, and is provided with documented purity and recommended storage conditions for safe handling.
Catalog No. 50-002-82876