Learn More
Medchemexpress LLC 2,4-Dichlorobenzenesulfonyl chloride | 16271-33-3 | ≥98.0% | 245.51 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW013409100G
Encompass_Preferred
2,4-Dichlorobenzenesulfonyl chloride is a heterocyclic building block used in the synthesis of nicotinyl arylsulfonylhydrazides. These synthesized compounds show potential anticancer and antibacterial activities, making this product suitable for various research applications.
  • Can be used in the synthesis of nicotinyl arylsulfonylhydrazides
  • Exhibits potential anticancer activities
  • Exhibits potential antibacterial activities
  • For research use only
Catalog No. 50-004-54273