Learn More
Medchemexpress LLC 2,4-dichlorobenzyl alcohol | 1777-82-8 | MFCD00004606 | 177.03 g/mol | C7H6Cl2O | 10 G
SDP
Supplier:  Medchemexpress LLC HYW03945410G
Encompass_Preferred
2,4-Dichlorobenzyl alcohol is a benzyl alcohol derivative used as a mild antiseptic and research chemical. It is provided for laboratory applications such as formulation, solubility studies, and analytical reference standards. The compound is soluble in DMSO and compatible with common in vivo vehicles; handle and store under recommended conditions. For research use only.
  • Used as a mild antiseptic and topical antimicrobial agent.
  • Suitable as an analytical reference standard for testing and quality control.
  • High DMSO solubility: 100 mg/mL (may require ultrasonic).
  • Compatible with in vivo formulation vehicles achieving ≥2.5 mg/mL.
  • Stable when stored as powder or in solvent under recommended temperatures.
Catalog No. 50-003-00533