Learn More
Medchemexpress LLC 2,4-dichlorobenzyl alcohol | 1777-82-8 | MFCD00004606 | 177.03 g·mol⁻¹ | C7H6Cl2O | 100 G
SDP
Supplier:  Medchemexpress LLC HYW039454100G
Encompass_Preferred
2,4-Dichlorobenzyl alcohol is a benzyl alcohol derivative with antiseptic properties commonly used as a research standard and as an active constituent in oral antiseptic formulations. It is supplied as a solid powder suitable for analytical testing, formulation studies, and other laboratory applications.
  • Antiseptic activity against a broad range of bacteria and some viruses.
  • Suitable as an analytical or reference standard for method development.
  • Powder form enables accurate weighing and formulation.
  • Well-characterized chemical identifiers: CAS 1777-82-8; formula C7H6Cl2O; molecular weight 177.03 g·mol⁻¹.
Catalog No. 50-002-99817