Learn More
Medchemexpress LLC 2,4-Dichlorobenzyl alcohol | 1777-82-8 | MFCD00004606 | 99.0% | 177.03 | C7H6Cl2O | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW039454
Encompass_Preferred
2,4-Dichlorobenzyl alcohol is an antiseptic aromatic alcohol (C7H6Cl2O, MW 177.03) supplied for use as an analytical reference standard and research reagent. It is used in qualitative and quantitative laboratory methods and method development, and requires appropriate handling per the SDS due to reported prenatal developmental toxicity.
  • Analytical reference standard suitable for HPLC, GC, and MS.
  • Orally active antiseptic used in antimicrobial and antiviral research.
  • Available in small standard and bulk solid forms for method development and bulk use.
  • Appropriate for qualitative and quantitative method validation and calibration.
  • Requires standard laboratory handling and safety precautions per SDS.
Catalog No. 50-003-04026