Learn More
Medchemexpress LLC 2,4-dichlorobenzyl alcohol | 1777-82-8 | MFCD00004606 | 99.0% | 177.03 g·mol⁻¹ | C7H6Cl2O | 1 ML

Supplier: Medchemexpress LLC HYW03945410MM/1ML
2,4-dichlorobenzyl alcohol is a small-molecule antiseptic (CAS 1777-82-8) used in research and analytical applications. Provided in small-volume, pre-made 10 mM solutions suitable for assay development and method validation. Refer to the supplier datasheet and SDS for handling, storage, and safety information.
- Mild antiseptic activity against bacteria and some viruses.
- Supplied as a 10 mM solution for convenient use.
- Small 1 mL volume suitable for analytical assays.
- Molecular weight 177.03 g·mol⁻¹ for stoichiometric calculations.
- Molecular formula C7H6Cl2O for identity confirmation.
- Datasheet and SDS available for safety and handling.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.