Learn More
Medchemexpress LLC 2,4-dichlorobenzyl alcohol | 1777-82-8 | MFCD00004606 | 99.8% | 177.03 g/mol | C7H6Cl2O | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0394545G
Encompass_Preferred
2,4-dichlorobenzyl alcohol is an antiseptic benzyl alcohol derivative used in research for its antibacterial and antiviral activity. It is supplied as a high-purity analytical standard in solid and solution forms for laboratory applications such as antiseptic testing, formulation studies, and analytical reference.
  • High purity (99.84%) for analytical applications.
  • Suitable as an analytical standard and research reagent.
  • Available as solid and as solution in DMSO for immediate use.
  • Solid form offers stable storage and ease of formulation.
  • Supplied with a safety data sheet for safe handling and regulatory compliance.
Catalog No. 50-002-99762