Learn More
Medchemexpress LLC 2,4-dichlorobenzyl alcohol | 1777-82-8 | MFCD00004606 | 99.8% | 177.03 g/mol | C7H6Cl2O | 50 G

Supplier: Medchemexpress LLC HYW03945450G
2,4-Dichlorobenzyl alcohol is a benzyl alcohol derivative used as a mild oral and throat antiseptic and as a research standard. It exhibits antibacterial and antiviral activity relevant to mouth and throat infections and has been employed in studies involving coronaviruses. Handle as a research chemical; animal data indicate potential prenatal developmental toxicity with certain exposures.
- Cas number: 1777-82-8.
- Chemical formula: C7H6Cl2O.
- Molecular weight: 177.03 g/mol.
- Purity: 99.84%.
- Typical packaging: 50 g.
- Storage: powder -20 °C (3 years) or 4 °C (2 years); in solvent -80 °C (6 months) or -20 °C (1 month).
- Applications: mild oral/throat antiseptic and research reagent.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.