Learn More
Medchemexpress LLC 2,4-Dichlorophenoxyacetic acid sodium | 2702-72-9 | 97% | 243.02 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY18572AR
Encompass_Preferred
2,4-D sodium (Standard) is the analytical standard of 2,4-D sodium (HY-18572A), intended for research and analytical applications. It is a selective herbicide that can induce apoptosis and inhibits DNA and protein synthesis, preventing normal plant growth and development.
  • Selective herbicide for broad-leaved weeds
  • Induces apoptosis
  • Inhibits DNA and protein synthesis
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-003-83910