Learn More
Medchemexpress LLC 2- 4-Diethylamino-2- 500g
SDP
Supplier:  Medchemexpress LLC HYW099393500G
Encompass_Preferred
2-(4-Diethylamino-2-hydroxybenzoyl)benzoicacid is an intermediate in the synthesis of rhodamine derivatives[1]
Catalog No. 50-002-05591