Learn More
Medchemexpress LLC 2,4-Difluorobenzoic acid-d3 | 1219804-63-3 | 161.12 | 1 MG

Supplier: Medchemexpress LLC HYW002068S1MG
This product is 2,4-Difluorobenzoic acid-d3, supplied by MedChemExpress as a white to off-white solid. It is designed for laboratory research and the manufacture of various substances. With a purity of 99% (HPLC) and an isotopic enrichment of 98.6%, this chemical ensures reliability for scientific applications. Stable under recommended storage conditions, it provides consistent performance.
- High purity of 99% (HPLC)
- Isotopic enrichment of 98.6%
- Supplied as a solid for ease of handling
- Suitable for laboratory use
- Stable for long-term storage as powder at -20°C for 3 years
- Stable in solvent for up to 6 months at -80°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.