Learn More
Medchemexpress LLC 2,4-Difluorobenzoic acid-d3 | 1219804-63-3 | 161.12 | 5 MG

Supplier: Medchemexpress LLC HYW002068S5MG
2,4-Difluorobenzoic acid-d3 is a chemical substance primarily used for laboratory research and manufacturing. This compound is a deuterated form of 2,4-Difluorobenzoic acid.
- Appears as a white to off-white solid
- Purity of 99% as determined by HPLC
- Molecular formula: C7HD3F2O2
- Molecular weight: 161.12
- Powder storage at -20°C for 3 years
- Storage in solvent at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.