Learn More
AdipoGen 2 4-Difluororesorcinol
SDP
Supplier:  AdipoGen CDXD0329G005
Encompass_Preferred
Chemical. CAS 195136-71-1. Formula C6H4F2O2. MW 146.09. Synthetic Fluorinated building block for dye synthesis. Key precusor for preparing Pacific Blue dyes. Precursor that can be used for preparing fluorescent dyes such as fluorescein, coumarin and resorufin analogs.
Catalog No. 50-256-1119