Learn More
Medchemexpress LLC 2′,4′-dihydroxyacetophenone | 89-84-9 | MFCD00002279 | 100.0% | 152.15 g/mol | C8H8O3 | 500mg
SDP
Supplier:  Medchemexpress LLC HYY0694500MG
Encompass_Preferred
2 4 -Dihydroxyacetophenone (Resacetophenone) is acetophenone carrying hydroxy substituents at positions 2 and 4 2 4 -Dihydroxyacetophenone involves in a practical CsHCO3-mediated alkylation that efficiently provide 4-alkylated products with excellent regioselectivity good isolated yields and a broad substrate scope 2 4 -Dihydroxyacetophenone is a plant metabolite[1][2][3]
Catalog No. 50-002-67880