Learn More
Medchemexpress LLC 2,4-dihydroxybenzophenone-13C6 | 2731164-01-3 | 220.17 g·mol⁻1 | C713C6H10O3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW014223S1MG
Encompass_Preferred
2,4-dihydroxybenzophenone-13C6 is a 13C6-labeled isotopically enriched form of 2,4-dihydroxybenzophenone used as an ultraviolet absorber and as an analytical internal standard or tracer for quantitative techniques such as NMR, GC-MS, and LC-MS. It is supplied as a research reagent for analytical applications (CAS 2731164-01-3; MW 220.17).
  • isotopically labeled (13C6) for clear mass spectrometric distinction and quantitation.
  • formulated for use as an internal standard in NMR, GC-MS, and LC-MS workflows.
  • suitable for tracer studies and quantitative analysis.
  • provided as a stable powder for long-term storage under recommended conditions.
Catalog No. 50-003-04037