Learn More
Medchemexpress LLC 2,4-dihydroxypyridine | 626-03-9 | MFCD00006273 | 100.0% | 111.10 g/mol | C5H5NO2 | 100g
SDP
Supplier:  Medchemexpress LLC HYY0426100G
Encompass_Preferred
2 4-Dihydroxypyridine (compound 2) is a pyridine derivative which is active against both Topo IIKHIV activity and HIV-1 replication with a pIC50 of 5 05 and 4 07 against Topo IIKHIV and HIV-1 replication reespectively[1]
Catalog No. 50-002-68464