Learn More
Medchemexpress LLC 2,4-Dimethoxybenzyl alcohol | 7314-44-5 | 168.19 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW007584
Encompass_Preferred
2,4-Dimethoxybenzyl alcohol is an aromatic alcohol that serves as a substrate for glucose-methanol-choline (GMC) oxidoreductase. GMC oxidoreductase is characterized as an aryl-alcohol oxidase. This compound also functions as a drug intermediate.
  • Aromatic alcohol
  • Substrate for glucose-methanol-choline (GMC) oxidoreductase
  • Functions as a drug intermediate
  • Appearance is solid below 35°C and liquid above 39°C
  • Color ranges from white to off-white
Catalog No. 50-004-48024