Learn More
Medchemexpress LLC 2,4-Dimethoxybenzyl alcohol | 7314-44-5 | 4614 | 168.19 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00758410G
Encompass_Preferred
2,4-Dimethoxybenzyl alcohol is an aromatic alcohol and a substrate of glucose-methanol-choline (GMC) oxidoreductase, which exhibits characteristics of an aryl-alcohol oxidase.
  • Can be used as a drug intermediate
  • Substrate of glucose-methanol-choline (GMC) oxidoreductase
  • Displays characteristics of an aryl-alcohol oxidase
Catalog No. 50-004-45647