Learn More
Medchemexpress LLC 2-(4-Fluorophenyl)acetic acid-d2 | 113715-48-3 | 156.15 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW001955S5MG
Encompass_Preferred
2-(4-Fluorophenyl)acetic acid-d2 is the deuterium labeled 2-(4-Fluorophenyl)acetic acid. This compound is intended for research use only and can serve as a tracer or an internal standard for quantitative analysis. Deuterium substitution has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotope for drug development research
Catalog No. 50-004-45557