Learn More
Medchemexpress LLC 2-(4-fluorophenyl)thiazolo[5,4-b]pyridin-3-one | 158093-65-3 | MFCD13189577 | 99.5% | 246.26 g·mol⁻¹ | C12H7FN2OS | 10MG
SDP
Supplier:  Medchemexpress LLC HY12469610MG
Encompass_Preferred
PU139 is a potent pan-histone acetyltransferase (HAT) inhibitor used in research to probe histone acetylation and chromatin regulation. It inhibits multiple HAT enzymes including Gcn5, PCAF, CBP, and p300 with reported IC50 values in the low micromolar range. The compound is supplied as a solid or as a DMSO solution, and is provided at high purity for laboratory research applications.
  • Potent pan-HAT inhibitor with low micromolar IC50s.
  • Useful for biochemical and cell-based studies of histone acetylation.
  • High purity suitable for research applications.
  • Available as solid and pre-made DMSO solution.
  • Stable when stored sealed and kept away from moisture.
Catalog No. 50-002-37191