Learn More
Medchemexpress LLC 2-(4-formylphenyl)propionic acid | 43153-07-7 | MFCD08275588 | C10H10O3 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13126010MG
Encompass_Preferred
Ibuprofen Impurity K (2-(4-formylphenyl)propionic acid; CAS 43153-07-7) is a known degradation product of ibuprofen used as an analytical reference standard in impurity profiling, stability testing, and method development. It is a racemic aromatic propionic acid (C10H10O3, MW 178.18) typically supplied in milligram quantities for laboratory and quality-control use.
  • Used as an analytical reference standard for impurity identification and quantification.
  • Applicable to stability testing and forced-degradation studies.
  • Suitable for HPLC method development and validation.
  • Provided as a racemic form for representative impurity profiling.
  • Molecular formula C10H10O3 and molecular weight 178.18 for analytical calculations.
  • Available in small quantities suitable for research and quality-control applications.
Catalog No. 50-002-61175