Learn More
Medchemexpress LLC 2-[4-(hydroxymethyl)phenoxy]acetic acid | 68858-21-9 | MFCD00057827 | 98.3% | 182.18 g/mol | C9H10O4 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0164131G
Encompass_Preferred
4-(Hydroxymethyl)phenoxyacetic acid (CAS 68858-21-9) is an acid-labile resin linkage agent intended for solid-phase peptide synthesis (SPPS). Supplied as a white to off-white solid with high purity, it facilitates controlled cleavage of peptide chains from resin under acidic conditions and is provided for research use.
  • Acid-labile linkage for controlled cleavage under acidic conditions.
  • Compatible with common solid-phase peptide synthesis chemistries.
  • High purity suitable for research applications.
  • White to off-white solid for easy handling and storage.
  • Enables efficient release of synthesized peptides from resin.
  • Well characterized (C9H10O4, 182.18 g/mol).
Catalog No. 50-003-02162