Learn More
Medchemexpress LLC 2-(4-hydroxyphenyl)acetonitrile | 14191-95-8 | MFCD00002383 | 98.0% | 133.15 g/mol | C8H7NO | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0158235G
Encompass_Preferred
4-Hydroxybenzyl cyanide (2-(4-hydroxyphenyl)acetonitrile; CAS 14191-95-8) is a small organic compound and endogenous metabolite used as a reagent, reference standard, and synthetic intermediate in biochemical and analytical research. It is supplied at high purity and is suitable for analytical, metabolic, and synthetic applications.
  • High listed purity (98.0%) suitable for analytical work.
  • Useful as a reference standard and synthetic intermediate.
  • Available as a solid and as a 10 mM solution in DMSO.
  • Molecular formula C8H7NO; molecular weight 133.15 g/mol.
  • Stable when stored according to the safety data sheet recommendations.
Catalog No. 50-003-00527
 Restricted Item