Learn More
Medchemexpress LLC 2-(4-hydroxyphenyl)benzo[b]thiophene-6-ol | 63676-22-2 | MFCD16619490 | 99.9% | 242.29 g/mol | C14H10O2S | 5 MG

Supplier: Medchemexpress LLC HY1102015MG
Estrogen receptor modulator 1 is a small-molecule research compound (CAS 63676-22-2) used in in vitro and in vivo studies of estrogen receptor activity. It has formula C14H10O2S and a molecular weight of 242.29 g/mol; supplied as a white to off-white solid with high reported purity.
- Used for estrogen receptor modulation studies
- Molecular formula C14H10O2S
- Molecular weight 242.29 g/mol
- Supplied as a white to off-white solid
- High purity (≈99.9%)
- Available in small research pack sizes
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.