Learn More
Medchemexpress LLC 2,4-Imidazolidinedione, 5-hydroxy-1-methyl- | 84210-26-4 | 98.0% | 130.10 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11131450MG
Encompass_Preferred
NZ 419 (5-Hydroxy-1-methylhydantoin) is a metabolite of creatinine and an antioxidant with oral activity, which has the ability to scavenge hydroxyl radicals (•OH). It can inhibit the progression of chronic kidney disease (CKD) in rats.
  • Metabolite of creatinine
  • Antioxidant with oral activity
  • Scavenges hydroxyl radicals (•OH)
  • Inhibits progression of chronic kidney disease in rats
Catalog No. 50-003-43106