Learn More
Medchemexpress LLC 2,4-imidazolidinedione, 5-phenyl- | 89-24-7 | ≥97.0% | 176.18 | C9H8N2O2 | 50MG

Supplier: Medchemexpress LLC HY12578050MG
5-Phenylhydantoin is an antiepileptic small molecule provided as a research reference standard. It has reported hypolipidemic activity and is supplied as a powder for biochemical and pharmacological studies. This product is intended for laboratory research use only and is not for human or clinical applications.
- High chemical purity: ≥97.0%.
- Defined molecular formula: C9H8N2O2, molecular weight 176.18.
- Supplied as a powder suitable for analytical and preparative work.
- Common package sizes available: 25 mg, 50 mg, 100 mg; larger quantities available on request.
- Storage recommendations: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).
- Intended use: research applications and reference standard.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.