Learn More
Medchemexpress LLC 2-(4-methylpiperazin-1-yl)-5-(2,3,5-trichlorophenyl)pyrimidin-4-amine | 130800-90-7 | 99.6% | 372.68 | C15H16Cl3N5 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10833510MM 1ML
Encompass_Preferred
Sipatrigine is a neuroprotective small molecule that inhibits glutamate release and blocks voltage-dependent sodium and calcium channels. Supplied as a ready-to-use 10 mM solution in DMSO, this reagent is intended for research applications such as in vitro pharmacology and electrophysiology studies.
  • Ready-to-use 10 mM solution in DMSO.
  • Convenient 1 mL volume for small-scale experiments.
  • High purity (99.6%) suitable for research use.
  • Molecular weight 372.68; CAS 130800-90-7.
  • Reported activity as sodium and calcium channel inhibitor and glutamate release inhibitor.
Catalog No. 50-002-43343