Learn More
Medchemexpress LLC 2-(4-nitrophenyl)imidazo[2,1-b]-5,6,7,8-tetrahydrobenzothiazole | 60477-38-5 | MFCD05670249 | 99.4% | 299.35 | C15H13N3O2S | 10MG
SDP
Supplier:  Medchemexpress LLC HY12307610MG
Encompass_Preferred
Pifithrin-α, p-nitro, cyclic is a cell-permeable inhibitor of p53 transcriptional activity used in research to modulate the MDM-2/p53 pathway. It is a p-nitro substituted cyclic analog of pifithrin-α with greater potency and a longer half-life, molecular formula C15H13N3O2S, molecular weight 299.35, and CAS 60477-38-5. Supplied in small milligram quantities for in vitro dose-response studies.
  • Cell-permeable p53 inhibitor for in vitro studies.
  • Greater potency and longer half-life than pifithrin-α.
  • Suitable for studies targeting the MDM-2/p53 pathway.
  • Available in small milligram quantities for dose-response experiments.
  • High reported purity for reliable experimental results.
Catalog No. 50-002-39119